Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-serine Mouse anti-Vertebrates, Clone: 4A4, MilliporeSigma™

Catalog No. 051000XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-100-0XMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-100-0XMI Supplier MilliporeSigma Supplier No. 051000X
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phosphoserine Antibody, clone 4A4 is an antibody against Phosphoserine for use in western blot, immunofluoresence, flow cytometry, immunohistochemistry (paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4A4
Conjugate Unconjugated
Formulation 50μL of PBS with 0.05% sodium azide with 30% glycerol.
Host Species Mouse
Immunogen Phosphoserine coupled to KLH
Purification Method Protein G-Sepharose™ chromatography
Quantity 50 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2 years at -20°C from date of shipment. For maximum recovery of the product, centrifuge the original vial prior to removing the cap.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.