Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase Rabbit anti-Human, Monkey, Canine, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 09482MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-482-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-482-MI Supplier MilliporeSigma Supplier No. 09482
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PI3 Kinase Antibody, p110β is an antibody against PI3 Kinase for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase, p110β
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Antigen affinity purified rabbit serum in 0.1M Tris-Glycine (pH7.4), 150mM NaCl, with 0.05% NaN3.
Gene Symbols EC 2.7.1.153 ; MGC142161; MGC142163; PI3K; p110-alpha
Host Species Rabbit
Immunogen KLH-conjugated synthetic linear peptide corresponding to the internal domain of PI3 Kinase, p110β.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_006210.1
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.