Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3-Kinase p170 Mouse, Unlabeled, Clone: 17, BD

Catalog No. BDB611046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611046 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611046 Supplier BD Biosciences Supplier No. 611046
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), producing PtdIns(3)P, PtdIns(3,4)P2, and PtdIns(3,4,5)P3. PI3-Kinase is a heterodimer of an 85kDa regulatory subunit (p85) and a 110kDa catalytic subunit (p110). However, this heterodimer is only one member of a larger family of proteins with similarity to the p110 subunit. These different PI3-kinase isoforms have been divided into three classes. Class I consists of p110α and p110β which bind the p85 subunit and associate with receptor tyrosine kinases. Class II contains 68D, cpk, and p170. These proteins all contain a C-terminal C2 domain and phosphorylate PtdIns and PtdIns(4)P, but not
PtdIns(4,5)P2. Class III members only phosphorylate PtdIns to PtdIns(3)P and include the S. cerevisiae Vps34p and its human homologs. The first human Class II PI3-kinase to be described, p170, contains the characteristic C-terminal phosphoinositide-binding C2 domain also found in PKC isoforms and synaptotagmins. In addition, expression of PI3-kinase p170 is widespread and may indicate that the protein performs a housekeeping function in cell growth and differentiation.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3-Kinase p170
Applications Western Blot
Classification Monoclonal
Clone 17
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse PI3-Kinase p170
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.