Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI3 Kinase p85 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABS233MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS233MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS233MI Supplier MilliporeSigma Supplier No. ABS233
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PI3 Kinase regulatory subunit alpha using this rabbit polyclonal Anti-PI3 Kinase p85, N-SH2 domain, Cat. No. ABS233, validated for use in Immunoprecipitation and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3 Kinase p85
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols Pik3r1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the N-SH2 domain of rat PI3 Kinase p85.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_037137
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.