Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4-Kinase β Mouse, Unlabeled, Clone: 7, BD

Catalog No. BDB611816 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611816 50 μg
BDB611817 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611816 Supplier BD Biosciences Supplier No. 611816
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphoinositide turnover is a well established mechanism of intracellular signal transduction. Sequential phosphorylation of phosphatidylinositol (PtdIns) results inPtdIns-4-phosphate (PIP) and PtdIns-4,5-bisphosphate (PIP2). Phospholipase C (PLC)hydrolyzes PIP2 to inositol-1,4,5-trisphosphate (IP3) which stimulates release of intracellular Ca2+. PIP is generated by phosphorylation of PtdIns at the D4 position of the inositol ring. This event is mediated by the PtdIns 4-kinases (PI4-K). These enzymes are divided into two types (II and III) based on their size and sensitivity to certain compounds. Although the PI4-Ks are abundantly distributed throughout the cell, activity is found primarily in association with membranous structures. Members of this family contain a lipid kinase unique domain and a C-terminal catalytic domain. Two mammalian PI4-Ks, PI4-Kα and PI4-Kβ, have been identified. PI4-Kβ is homologous to the yeast PI4-K, PIK1. Based on its size and sensitivity to wortmanin (a PI3-K inhibitor), PI4-Kβ is classified as a type III enzyme. Although it is found in the cytosol and in association with the Golgi, the specific function of PI4-Kβ is yet to be determined.

Host Species: Mouse
Clone: 7
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human PI4Kβ aa. 411-626

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4-Kinase β
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PI4Kβ aa. 411-626
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.