Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PIK3R1, HRAS Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89023314 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-314 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-314 Supplier Abnova Corporation Supplier No. DI0555
Add to Cart
Add to Cart

Includes

PIK3R1 rabbit purified polyclonal antibody (20μg); HRAS mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PIK3R1 protein, and the other against the HRAS protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen PIK3R1
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene PIK3R1
Gene Alias GRB1/p85/p85-ALPHA
Gene Symbols PIK3R1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5295/3265
Includes Antibody pair set content: 1. PIK3R1 rabbit purified polyclonal antibody (20 ug) 2. HRAS mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type PIK3R1 & HRAS Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.