Learn More
ECM Bioscience PKCδ (N-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM2421
Encompass_Preferred
Clone (M242) was generated from a recombinant human protein that included amino acids residues in the N-terminal region. This sequence is conserved in rat and mouse PKCδ, and has low homology to other PKC family members. Accession #Q05655. This antibody detects a 78 kDa* protein corresponding to the molecular mass of PKCδ on SDS-PAGE immunoblots of adult mouse brain tissue lysate. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PKCdelta
Catalog No. 50-206-8796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.