Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PNMA2 Ab-1 Matched Antibody Pair, Abnova™

Numéro de catalogue. 89023642 missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89-023-642 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89-023-642 Fournisseur Abnova Corporation Code fournisseur H00010687AP41
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Comprend

Capture antibody: mouse monoclonal anti-PNMA2 (100μg); Detection antibody: biotinylated mouse monoclonal anti-PNMA2, IgG2a Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PNMA2

Host Species: Mouse, Mouse
Isotype: IgG, IgG2a κ
Species Reactivity: Hu

ELISA Pair

Spécifications

Antigen PNMA2
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PNMA2.
Gene PNMA2
Gene Alias KIAA0883/MA2/MM2/RGAG2
Gene Symbols PNMA2
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10687
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-PNMA2 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-PNMA2, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Isotype IgG, IgG2a κ
Afficher plus de résultats Afficher moins de résultats
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.