Learn More
ABclonal Technology POLI Polyclonal Antibody, 100UL, Validated in:WB, IHC
Supplier: ABclonal Technology A1942
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 50-350 of human POLI (NP_009126.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB•Swiss Prot: Q9UNA4•Gene ID:11201•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.