Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly ADP-ribose Mouse anti-Human, Clone: 10H, MilliporeSigma™

Catalog No. MABC547MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC547MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC547MI Supplier MilliporeSigma Supplier No. MABC547
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Detect PARP using this mouse monoclonal antibody, Anti-Poly ADP-ribose Antibody, clone 10H validated for use in western blotting, immunoprecipitation and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Poly ADP-ribose
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10H
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Corresponding to human Poly ADP-ribose chain.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Product Type Antibody
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.