Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyglutamine-Expansion Diseases Marker Mouse anti-Human, Clone: 5TF1-1C2, MilliporeSigma™

Catalog No. MAB1574MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1574MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1574MI Supplier MilliporeSigma Supplier No. MAB1574
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2 is an antibody against Polyglutamine-Expansion Diseases Marker for use in ELISA, immunocytochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polyglutamine-Expansion Diseases Marker
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5TF1-1C2
Conjugate Unconjugated
Formulation Ascites fluid containing no preservatives.
Host Species Mouse
Immunogen N-terminal part of the human TATA Box Binding Protein (TBP).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.