Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysialic Acid-NCAM Mouse anti-Human, Rat, Mouse, Vertebrates, Clone: 2-2B, MilliporeSigma™

Catalog No. MAB5324MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5324MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5324MI Supplier MilliporeSigma Supplier No. MAB5324
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Polysialic Acid-NCAM using this Anti-Polysialic Acid-NCAM Antibody, clone 2-2B validated for use in immunocytochemistry, immunohistochemistry, RIA and western blotting with more than 30 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polysialic Acid-NCAM
Applications Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 2-2B
Conjugate Unconjugated
Formulation Mouse monoclonal IgM in buffer containing 50% glycerol, no preservatives.
Gene Accession No. NM_000615.5
Gene Symbols NCAM1, CD56, NCAM-140, NCAM, NCAM-120, MSK39
Host Species Mouse
Immunogen Viable Meningococcus group B (strain 355).
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.