Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-Prion Protein antibody, Mouse monoclonal
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P0110200UL
Encompass
Anti-Prion Protein antibody, Mouse monoclonal | clone 8H4, ~1.5 mg/mL, unconjugated, species reactivity: bovine, mouse, monkey, human, sheep, rat, 200uL
Catalog No. NC2737139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.