Learn More
Abcam Anti-Pro-Collagen III Alpha 1 Antibody, Rabbit Multiclonal, 1 mL
Description
- Host Species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Applications: Flow Cyt (Intra), ICC/IF, IHC-Fr, IHC-P
- Species Reactivity: Human, Mouse, Rat
- Purification Technique: Affinity purification Protein A
- Form: Liquid
- Storage Buffer: pH 7.2 to 7.4, 0.01% Sodium azide, PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Pro-Collagen III alpha 1 (Human, Mouse, Rat) |
| Buffer | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH7.2 to 7.4 |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Cross Reactivity | Human, Mouse, Rat |
| Formulation | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH 7.2 to 7.4 |
| Quantity | 1 mL |
| For Use With (Application) | Flow Cytometry (Intracellular), ICC/IF, IHC-Fr, IHC-P |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.