Learn More
Advanced ImmunoChemical, Inc. ANTI-PROCALCITONIN MAB
Supplier:  Advanced ImmunoChemical, Inc. 2PCAL1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs in vivo:6F10, 38F11, 44D9, 22A11, 18B7Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cellsof Balb/c mice immunized with fragments of human PCT conjugated with carrier protein.MAbs in vitro:27A3cc, 42cc, 14C12cc, P123, P124, P135, P160Specificity:MAb Isotypes:IgG1 for MAbs 6F10, 38F11, 22A11, 14C12cc, 18B7, P123, P124, P160IgG2a for MAbs 27A3cc, 42cc, 44D9, P135 Applications:MAbs 27A3cc, 42cc, 14C12cc, 44D9, 6F10, 38F11, 22A11, 18B7 are working in Western blotting.Purification:Protein A chromatographyPresentation:PBS, pH 7.4, 0.09 % sodium azide (NaN₃)Storage Conditions:+4 °C (+2 … +8 °C allowed)
Catalog No. NC3566225