Learn More
Sartorius Biosensor / Protein G (ProG) Tray
Supplier:  Sartorius 185082
Encompass
Protein G (ProG) Biosensors - One tray of 96 biosensors coated with Protein G. For quantitation applications.
Catalog No. NC9070309
Add to Cart
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov