Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B Mouse anti-Human, Clone: FG6-1G, MilliporeSigma™

Catalog No. MABS197MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS197MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS197MI Supplier MilliporeSigma Supplier No. MABS197
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PTP1B Antibody, clone FG6-1G is an antibody against PTP1B for use in Western Blotting, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP1B
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone FG6-1G
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002818
Gene Symbols PTPN1, PTP1B
Host Species Mouse
Immunogen Recombinant protein corresponding to the catalytic domain of human PTP1B.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.