Learn More
ECM Bioscience PTP1B (Tyr-152), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP2391
Encompass_Preferred
This antibody was cross-adsorbed to a phospho-tyrosine peptide then affinity purified with phospho-PTP1B (Tyr-152) peptide. The purified antibody detects a 50 kDa* band corresponding to PTP1B in western blots of pervanadate treated human A431 and Jurkat, as well as mouse SYF cells. This reactivity is not observed in control cells or after alkaline phosphatase treatment of blots. The immunogen was PTP1B (Tyr-152) synthetic peptide corresponds to amino acids surrounding Tyr-152 in human PTP1B. This sequence has significant homology to the conserved site in rat and mouse PTP1B, and has homology to conserved sites in TC-PTP and MPTP, but not other protein tyrosine phosphatase family members. Accession #P18031. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PTPN1; TCPTP/PTPN2
Catalog No. 50-206-8808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.