Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein tyrosine phosphatase domain containing 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-617 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-617 Supplier Abnova Corporation Supplier No. H00138639B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PTPDC1 protein.

The protein encoded by this gene contains a characteristic motif of protein tyrosine phosphatases (PTPs). PTPs regulate activities of phosphoproteins through dephosphorylation. They are signaling molecules involved in the regulation of a wide variety of biological processes. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding distinct isoforms have been identified. [provided by RefSeq

Sequence: MAAGVLPQNEQPYSTLVNNSECVANMKGNLERPTPKYTKVGERLRHVIPGHMACSMACGGRACKYENPARWSEQEQAIKGVYSSWVTDNILAMARPSSELLEKYHIIDQFLSHGIKTIINLQRPGEHASCGNPLEQESGFTYLPEAFMEAGIYFYNFGWKDYGVASLTTILDMVKVMTFALQEGKVAIHCHAGLGRTGVLIACYLVFATRMTADQAIIFVRAKRPNSIQTRGQLLCVREFTQFLTPLRNIFSCCDPKAHAVTLPQYLIRQRHLLHGYEARLLKHVPKIIHLVCKLLLDLAENRPVMMKDVSEGPGLSAEIEKTMSEMVTMQLDKELLRHDSDFSNPPNPTAVAADFDNRGMIFSNEQQFDPLWKRRNVECLQPLTHLKRRLSYSDSDLKRAENLLEQGETPQTVPAQILVGHKPRQQKLISHCYIPQSPEPDLHKEALVRSTLSFWSQSKFGGLEGLKDNGSPIFHGRIIPKEAQQSGAFSADVSGSHSPGEPVSPSFANVHKDPNPAHQQVSHCQCKTHGVGSPGSVRQNSRTPRSPLDCGSSPKAQFLVEHETQDSKDLSEAASHSALQSELSAEARRILAAKALANLNESVEKEELKRKVEMWQKELNSRDGAWERICGERDPFILCSLMWSWVEQLKEPVITKEDVDMLVDRRADAAEALFLLEKGQHQTILCVLHCIVNLQTIPVDVEEAFLAHAIKAFTKVNFDSENGPTVYNTLKKIFKHTLEEKRKMTKDGPKPGL

Specifications

Antigen protein tyrosine phosphatase domain containing 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PTPDC1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PTPDC1
Gene Accession No. BC067120.1
Gene Alias FLJ42922/PTP9Q22
Gene Symbols PTPDC1
Host Species Mouse
Immunogen PTPDC1 (AAH67120.1, 1 a.a. ∼ 754 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138639
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.