Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab11-FIP1/RCP Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABC513MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC513MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC513MI Supplier MilliporeSigma Supplier No. ABC513
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Rab11-FIP1/RCP Antibody is validated for use in western blotting, immunocytochemistry and immunoprecipitation for the detection of Rab11-FIP1/RCP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab11-FIP1/RCP
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.
Gene Symbols RAB11FIP1; RCP
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human Rab11-FIP1/RCP.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001002814
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.