Learn More
Rockland Immunochemicals Anti-RABBIT IgG (H&L) (GOAT) Antibody Peroxidase Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Ms Rt & Sh Serum Proteins) - 611-103-122
SDP
Supplier:  Rockland Immunochemicals 611103122
Encompass_Preferred
Goat anti-Rabbit IgG (H&L) Antibody Conjugated to Horseradish Peroxidase (HRP) is designed for chemiluminescent detection. Secondary antibodies bind to the primary antibody to assist in detection, sorting and purification of target antigens. To enable detection, the secondary antibody must have specificity for the antibody species and isotype of the primary antibody being used and generally is conjugated. This purified secondary antibody has been validated and optimized yielding good sensitivity and reproducible results with Rockland's primary antibodies. This anti-rabbit IgG antibody has been crossed adsorbed to show no reaction against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat and Sheep Serum Proteins.
Catalog No. NC0161557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.