Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF(TM) 488A antibody produced in donkey ~2 mg/mL, affinity isolated antibody | 250UL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SAB4600036250UL
Encompass
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF(TM) 488A antibody produced in donkey ~2 mg/mL, affinity isolated antibody | 250UL
Catalog No. NC1733720
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.