Learn More
Advanced ImmunoChemical, Inc. Anti-RABIES VIRUS MAB
Supplier:  Advanced ImmunoChemical, Inc. 5RVQT
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs in vitro:1C5ccMAbs in vivo:4G4, New MAbs: 4F1, 7E3Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified rabies virus. Specificity:MAbs 4G4, 4F1, 7E3 are specific to glycoprotein.MAb 1C5cc has a virus-neutralizing activity in respect to the majority of standard RV and rabies-relative viruses: CVS, Lagos bat, Mokola, Flury lep, Duvenhage and the field isolates.MAb Isotypes:IgG2a for MAbs 1C5ccIgG2b for MAbs 4G4, 4F1, 7E3Applications:MAbs 1C5cc, 4G4,4F1, 7E3 are working in EIA.MAbs 4F1, 7E3 are working in Western blotting.MAbs 4F1, 7E3 are working in immunofluorescence.MAb 1C5cc works in immunocytochemistry.Purification:MAb 1C5cc is purified by chromatography on protein G Sepharose and all other MAbs on protein A Sepharose.Presentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566232