Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Mouse anti-Human, Mouse, Rat, Clone: 23A8, MilliporeSigma™

Catalog No. 05389MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-389-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-389-MI Supplier MilliporeSigma Supplier No. 05389
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Rac1 Antibody, clone 23A8 has been published in more than 40 citations and validated for use in Immunohistochemistry (immunohistochemistry), Immunoprecipitation (IP) and Western Blotting (WB) for the detection of Rac1 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23A8
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_006908
Gene Symbols RAC1, MGC111543, ORFNames=MIG5, MIG5, TC-25, p21-Rac1
Host Species Mouse
Immunogen Recombinant protein containing the full length human Rac
Purification Method Protein G purfied
Quantity 250 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.