Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgG, Horseradish Peroxidase-Linked Species-Specific Whole Antibody (from goat) Secondary Antibody, Cytiva
GSA_VA

Catalog No. 45001200 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-200 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-200 Supplier Cytiva Supplier No. NA935
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use for protein blotting, ELISA and immunocytochemistry applications. Anti-Rat IgG, Horseradish Peroxidase-Linked Species-Specific Whole Antibody (from goat) Secondary Antibody, Cytiva, is a secondary antibody from goat.

Host Species: Goat
Isotype: IgG
Species Reactivity: Rt

Protein Blotting, ELISA, Immunocytochemistry

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Classification Monoclonal
Conjugate HRP
Host Species Goat
Purification Method Affinity Purified
Quantity 1 mL
Primary or Secondary Secondary
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.