Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Rat IgG2a Isotype Control, PE-Texas Red
Rat Monoclonal Isotype Control
Supplier: Invitrogen™ R2A17
Description
Rat IgG2a Isotype Control for Flow, Ctrl
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
Specifications
| Rat IgG2a | |
| Monoclonal | |
| PE-Texas Red | |
| IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
| Rat IgG2a | |
| 500 μL | |
| Isotype Control (Primary) | |
| 4°C, store in dark | |
| Liquid |
| Control, Flow Cytometry | |
| 100 μg/mL | |
| PBS with 4mg/mL BSA and 0.1% sodium azide | |
| Rat | |
| Purified | |
| RUO | |
| Not Applicable | |
| Isotype Control | |
| IgG2a |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction