Learn More
Advanced ImmunoChemical, Inc. ANTI-RAT MS INSULIN PROINS MAB
Supplier:  Advanced ImmunoChemical, Inc. 1IP1 MG
Encompass_Preferred
Monoclonal antibodies / Metabolic Syndrome / Monoclonal mouse anti-rat-mouse insulin/proinsulin. MAbs:D6C4, D3E7Specificity:MAbs D6C4 and D3E7 directed to different antigenic determinants. Kd for MAb D6C4 is 8.1 x 10-8 M.Kd for MAb D3E7 is 6.3 x 10-8 M.MAb D6C4 and D3E7 cross-react with human, bovine and porcine insulin and proinsulin.MAb Isotypes:IgG1, kappa for MAbs D6C4 and D3E7Applications:Detection of rat-mouse insulin/proinsulin in two-site enzyme immunoassay. Recommend pair (capture-detection): D6C4 – D3E7.Antibodies D6C4 and D3E7 can be used in immunohistochemistry on frozen sections.Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial Safety Note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. NC3565713