Learn More
Advanced ImmunoChemical, Inc. ANTI-RAT PROINSULIN MAB
Supplier:  Advanced ImmunoChemical, Inc. 1PINS1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragment of rat proinsulin conjugated with a carrier protein.Specificity:Rat proinsulinMAb Isotypes:IgG1 for MAb CCI-17Applications:MAb recognizes rat proinsulin in ELISA and Sandwich type immunoassay. MAb binds specifically to proinsulin and does not bind to insulin or C-peptide. See our “Rat C-peptide and proinsulin” Assay Notes for more information. For sandwich immunoassay it is recommended to use MAb in pair with MAbs specific to N-terminal part of C-peptides I and II (see also the data sheet for Cat.# 1-CP-r and “Rat C-peptide and proinsulin” Assay Notes for more information).The recommended pair for sandwich-type rat proinsulin I and II immunoassay is (capture- detection): CCI-17 – CII-55 Purification:Chromatography on protein A SepharosePresentation:Presentation: PBS, pH 7.4, 0.1 % sodium azide (NaN3)
Catalog No. NC3565724