Learn More
Description
This gene encodes a membrane protein that functions as part of a receptor complex for a small neuropeptide that increases intracellular cAMP levels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
Specifications
| Antigen | CGRP receptor component |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant RCP9. |
| Formulation | 50% glycerol |
| Gene | CRCP |
| Gene Accession No. | NM_014478 |
| Gene Alias | CGRP-RCP/MGC111194/RCP/RCP9 |
| Gene Symbols | CRCP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.