Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulator of G-protein signaling 19, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-485 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-485 Supplier Abnova Corporation Supplier No. H00010287A02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RGS19.

G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G-protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase-activating protein that functions to down-regulate Galpha i/Galpha q-linked signaling. Alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. [provided by RefSeq

Sequence: WNQERRRAWQASRESKLQPLPSCEVCATPSPEEVQSWAQSFDKLMHSPAGRSVFRAFLRTEYSEENMLFWLACEELKAEANQHVVDEKAR

Specifications

Antigen regulator of G-protein signaling 19
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RGS19.
Formulation 50% glycerol
Gene RGS19
Gene Accession No. NM_005873
Gene Alias GAIP/RGSGAIP
Gene Symbols RGS19
Host Species Mouse
Immunogen RGS19 (NP_005864, 51 a.a. ∼ 140 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10287
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.