Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Mouse anti-Human, Rat, Fish, Mouse, Clone: 4D2, MilliporeSigma™

Catalog No. MABN15MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN15MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN15MI Supplier MilliporeSigma Supplier No. MABN15
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rhodopsin Antibody, clone 4D2 is an antibody against Rhodopsin for use in immunohistochemistry, western blot, immunocytochemistry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4D2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_254276
Gene Symbols OPN2, RHO, RP4, CSNBAD, Opsin-2
Host Species Mouse
Immunogen Rat rod outer segments corresponding to rat Rhodopsin at the N-terminus.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Fish, Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.