Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RICTOR, RPS6KB1 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89023277 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-277 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-277 Supplier Abnova Corporation Supplier No. DI0518
Add to Cart
Add to Cart

Includes

RICTOR rabbit purified polyclonal antibody (20μg); RPS6KB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the RICTOR protein, and the other against the RPS6KB1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen RICTOR
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene RICTOR
Gene Alias DKFZp686B11164/KIAA1999/MGC39830/mAVO3
Gene Symbols RICTOR
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260/6198
Includes Antibody pair set content: 1. RICTOR rabbit purified polyclonal antibody (20 ug) 2. RPS6KB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type RICTOR & RPS6KB1 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.