Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rsk Mouse, Unlabeled, Clone: 78, BD

Catalog No. BDB610226 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610226 150 μg
BDB610225 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610226 Supplier BD Biosciences Supplier No. 610226
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The p90[rsk] (Rsk) and p70[s6k] kinases were first identified based on their ability to phosphorylate the 40S ribosomal protein S6 in vitro. Both of these enzymes are differentially regulated by serine/threonine phosphorylation in response to mitogenic stimulation. ERK1 and ERK2 have been shown to regulate Rsk activity. Once activated by this phosphorylation, a significant amount of Rsk can be found in the nucleus, suggesting that it has a role in nuclear signaling events. The regulation of nuclear Rsk and ERK activities by growth factors is coordinated with the induction of several early response genes. Rsk has also been shown to be activated by ionizing radiation, presumably through an activated MAP kinase. Studies in Xenopus oocytes and mouse NIH/3T3 cells indicate that inactive Rsk and ERK2 exist in a complex of approximately 110kDa. Upon phosphorylation of Rsk and ERK2, the heterodimer dissociates and at least a portion of these activated kinases translocate to the nucleus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rsk
Applications Western Blot
Classification Monoclonal
Clone 78
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.