Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX2 Mouse anti-Human, Clone: AS110, MilliporeSigma™

Catalog No. 051478MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-147-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-147-8MI Supplier MilliporeSigma Supplier No. 051478
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Anti-RUNX2 Antibody, clone AS110 detects level of RUNX2 and has been published and validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone AS110
Conjugate Unconjugated
Formulation Purified mouse monoclonal in 0.1M Tris-Glycine (pH 7.4) with 150mM NaCl and 0.05% NaN3.
Gene Accession No. NP_001019801
Gene Symbols AML3 , CBF-alpha-1, CBFA1, CCD, CCD1, MGC120022 , MGC120023, OSF-2, OSF2 , OTTHUMP00000016533, PEA2aA , PEBP2A , PEBP2A1, PEBP2A2 , PEBP2aA , PEBP2aA1
Host Species Mouse
Immunogen GST-coupled recombinant protein corresponding to amino acids 311-415 of human RUNX2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.