Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX3 Mouse, PE, Clone: R3-5G4, BD

Catalog No. BDB564814 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564814 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564814 Supplier BD Biosciences Supplier No. 564814
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R3-5G4 monoclonal antibody specifically binds to Runt-related transcription factor 3 (RUNX3) which is also known as Acute myeloid leukemia 2 protein (AML2), Core-binding factor subunit alpha-3 (CBFA3), and Polyomavirus enhancer-binding protein 2 alpha C subunit (PEBP2aC). RUNX3 is a member of the RUNX transcription factor family which includes RUNX1-3. RUNX3 is a key regulator of gene expression related to the development and differentiation of cells within the nervous and immune systems. In the immune system, RUNX3 is particularly involved in the commitment of CD8+ T cells in the thymus. RUNX3 is highly expressed and essential for the cytotoxic functions of NK cells, peripheral CD8+T cells and CD4+ CD8αα intraepithelial lymphocytes in the gut. RUNX3 als o plays a role in the differentiation and effector functions of Th1 cells . RUNX3 is activated downstream of the TGF-β signaling pathway and can play a role in tumor suppression. Aberrant expression of RUNX3 has been associated with tumorigenesis including the development of gastric and other cancers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX3
Applications Flow Cytometry
Classification Monoclonal
Clone R3-5G4
Conjugate PE
Description AML2; AML-2; CBFA3; PEBP2aC; PEBP2A3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RUNX3 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.