Learn More
S6 (pS235/pS236) Mouse, PE, Clone: N7-548, BD
Catalog No. BDB560433 Shop All BD Biosciences Products
Change view
Click to view available options
50 Tests; PE
Catalog No.
BDB560433 50 Tests; PE
1 options
This item is not returnable. View return policy
Catalog No. BDB560433 Supplier BD Biosciences Supplier No. 560433
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mouse Monoclonal Antibody
Ribosomal protein S6 (∽29kDa calculated and ∽32kDa observed molecular weights) is a component of the 40S ribosomal subunit and belongs to the S6E family of ribosomal proteins. The S6 ribosomal protein plays a role in regulating the translation of RNAs and thus controlling the growth and proliferation of cells. S6 ribosomal protein phosphorylation, especially at multiple C-terminal serine residues S235, S236, S240, and S244, activates S6. The activated S6 ribosomal protein in turn upregulates the ribosomal translation of RNA species coding for other ribosomal proteins, peptide elongation factors and other proteins involved in cell cycle entry and progression. These phosphorylations are mediated by various kinases (e.g., p70S6K and PKCD) activated through cellular responses to growth factors, cytokines, tumor promoting agents, and mitogens. The S6 ribosomal protein can be dephosphorylated in growth-arrested cells.