Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Serum Amyloid A (SAA), 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 2SAAH0.2MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal mouse anti-serum amyloid A (SAA). MAbs:SAA1, SAA6, SAA15,VSA6 ,VSA25Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant SAA, conjugated with carrier protein.Specificity:Human recombinant SAAMAb Isotypes:IgG1 for MAbs SAA1, SAA6, VSA6, VSA25 and SAA15Applications:SAA immunoassay. All MAbs recognize SAA in ELISA and Western blotting.Recommended pairs for sandwich immunoassay (capture-detection):SAA1 – SAA15SAA6 – SAA15SAA15 – SAA1SAA6 – SAA1SAA1 – SAA6VSA25 – VSA31VSA6 – VSA38Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °CMaterial Safety Note:This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9868