Learn More
Seracare Life Sciences Inc ANTI-SALMONELLA CSA-1 AB FITC
Supplier:  Seracare Life Sciences Inc 53300059
Encompass_Preferred
Affinity purified polyclonal antibody to the Salmonella Common Structural Antigens (CSA) made in Goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool of goats immunized with different strains of Salmonella.
Catalog No. 50-669-18
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.