Learn More
ACROBiosystems Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Human IgG1 (AM180)
Supplier:  ACROBiosystems SPDM180100UG
Encompass_Preferred
Antibody;100UG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.;Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Human IgG1 (AM128) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from human 293 cells (HEK293). This antibody recognizes the SARS-CoV-2 Spike Protein RBD domain and inhibits the interaction between SARS-CoV-2 RBD and ACE2 with an IC50 of 0.98 μg/mL using SARS-CoV-2 Inhibitor Screening Kit (Cat. No. EP-105).Pseudovirus assay shows that this antibody has potent neutralizing activity against pseudovirus bearing SARS-CoV-2 Spike protein.; ;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Human IgG1 (Cat. No. SPD-M180) with a linear range of 0.2-2 ng/mL (QC tested).;Serial dilutions of Anti-SARS-CoV-2 RBD Potent Neutralizing Antibo
Catalog No. 50-205-4820
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.