Learn More
ACROBiosystems ACROBiosystems Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122)
Supplier:  ACROBiosystems SPDM201100UG
Encompass_Preferred
Antibody;100UG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is detected with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122) (SPD-M201) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with Cynomolgus IgG1 constant domain. The mouse monoclonal antibody was obtained from a mouse immunized with recombinant SARS-CoV-2 Spike S1 Protein. ; The protein has a calculated MW of 48.8 kDa | 23.4 kDa. the protein migrates as 27 kDa and 55 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Spike RBD, His Tag (Cat. No. SPD-C52H1) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike RBD Neutralizing Antibo
Catalog No. 50-210-7033
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.