Learn More
ECM Bioscience Semaphorin-4D (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SM1881
Encompass_Preferred
Clone (M188) was generated from a recombinant protein corresponding to the C-terminal region of human Semaphorin-4D. Accession #Q92854 . This antibody detects a 150 kDa* band on SDS-PAGE immunoblots of adult mouse brain. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal, protein G purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names = CD100, Sema4D, SemaIVD
Catalog No. 50-206-8844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.