Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA5A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003108 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-108 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-108 Supplier Abnova Corporation Supplier No. H00009037A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SEMA5A.

Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).[supplied by OMIM

Sequence: PSFMEDNSRFSHVAVDVVQGREALVHIIYLATDYGTIKKVRVPLNQTSSSCLLEEIELFPERRREPIRSLQILHSQSVLFVGLREHVVKIPLKRCQFYRTRSTCIGA

Specifications

Antigen SEMA5A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SEMA5A.
Formulation 50% glycerol
Gene SEMA5A
Gene Accession No. NM_003966
Gene Alias FLJ12815/SEMAF/semF
Gene Symbols SEMA5A
Host Species Mouse
Immunogen SEMA5A (NP_003957, 393 a.a. ∼ 499 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9037
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.