Learn More
Market Lab Anti-Sera Vial Organizer Tray
Supplier:  Market Lab 9861
Encompass_Preferred
The Anti-Sera Vial Organizers from MarketLab are designed to organize and store anti-sera vials in the laboratory. They are made from durable polystyrene and feature removable dividers to accommodate different sized vials. The organizers have alphanumeric grid references for easy sample identification and retrieval. They are autoclavable and come in a variety of sizes to hold from 25 to 225 vials up to 20mm in diameter. The vial organizers help protect valuable samples while maximizing refrigerator and freezer space.
Catalog No. NC2830047
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.