Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPA1, Human, Matched Antibody Pair (AP44), Abnova™

Catalog No. 89301997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-301-997 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-301-997 Supplier Abnova Corporation Supplier No. H00653509AP44
Add to Cart
Add to Cart

Includes

Capture antibody: mouse monoclonal anti-SFTPA1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-SFTPA1, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

To detect and quantify protein level of human

Specifications

Antigen SFTPA1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SFTPA1.
Gene SFTPA1
Gene Alias COLEC4/FLJ51913/SFTP1/SP-A/SP-A1
Gene Symbols SFTPA1
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653509
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SFTPA1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SFTPA1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.