Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGLT-1 (Sodium Glucose Co-transporter-1) Rabbit anti-Canine, Human, Mouse, Pig, Rat, Rabbit, Feline, Avian, Reptile, Polyclonal, MilliporeSigma™

Catalog No. 071417MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-141-7MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-141-7MI Supplier MilliporeSigma Supplier No. 071417
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Anti-SGLT-1 (Sodium Glucose Co-transporter-1) Antibody detects level of SGLT-1 (Sodium Glucose Co-transporter-1) and has been published and validated for use in western blot, immunohistochemistry (paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGLT-1 (Sodium Glucose Co-transporter-1)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG antiserum in buffer containing 0.05% sodium azide.
Gene Symbols SGLT1; SLC5A1; NAGT; D22S675
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 402-420 in the cytoplasmic region of SGLT-1 of mouse/rabbit small intestine.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000343.1
Target Species Avian, Canine, Feline, Human, Mouse, Pig, Rabbit, Rat, Reptile
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.