Learn More
Description
Includes
Sheep Serum Blocking Reagent D, Avidin Blocking Reagent, Peroxidase Blocking Reagent,HSS-HRP (Vial B), Biotinylated Anti-sheep Antibody (Vial A), Biotin Blocking Reagent, AEC Chromogen Buffer, AEC Chromogen, Dropper Bottle
- Localization of antigens in broad range of histological and cytological specimens
- Detection is based on the formation of the Avidin-Biotin Complex (ABC) with primary antibodies
- Visualization is based on enzymatic conversion of a chromogenic substrate 3,3 Diaminobenzidine (DAB)
- Non-specific binding to phospholipids, nucleic acids and carbohydrate binding proteins
Applications: Immunohistochemistry
Conjugate: HRP- AEC
Storage: Store at 4°
Note: Kits are guaranteed for 6 months from date of receipt.
Specifications
Specifications
| Product Line | Novus |
| Product Type | Antibody DetectionKit |
| Quantity | 50 Tests |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
