Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Sialyl Lewis x Clone: KM93,

Catalog No. 565953100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
56-595-3100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-595-3100UG Supplier MilliporeSigma™ Supplier No. 565953100UG

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Sialyl Lewis x Clone: KM93 in Human samples, and it is validated for Blocking (Adhesion), Flow Cytometry, Immunohistochemistry (Paraffin)

Sialyl Lewis x [SLex; NeuAcα2,3Galβ1,4(Fucα1,3)GlcNAc] is a tetrasaccharide that acts as a minimal ligand (Kd = 129μM) for E-selectin (ELAM; endothelial leukocyte adhesion molecule), an endothelial cell-surface glycoprotein that mediates recruitment of neutrophils and monocytes to sites of endothelial inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sialyl Lewis x
Applications Inhibition Assays, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KM93
Conjugate Unconjugated
Formulation In PBS, 1% BSA, pH 7.2.
Host Species Mouse
Immunogen human lung adenocarcinoma cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the SLex carbohydrate epitope on glycoproteins and glycolipids on adenocarcinomas in lung primary tumors and with a variety of tumor cell lines.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.