Learn More
Valutek™ MacroTek Anti-Skid Polypropylene Shoe Covers
Description
- All Valutek™ shoe covers are tested and manufactured in ISO compliant facilities under individual inspection and strict process control to ensure Valutek™ quality standard and product specifications
- Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment
- Commonly used for protecting against dirt, grime, and certain dry particulates in nonhazardous environments
Specifications
Specifications
| Size | Large |
| Material | Polypropylene |
| Latex Free | No |
| Color | Blue |
| Disposable | Single-use |
| Closure Type | Elastic Ankle |
| Sole Type | Non-skid |
| Seam Type | Stitched |
| For Use With (Application) | Ideal for contamination control |
| Dimensions (L x W) | 16, 18 x 7 in. (40.64, 45.72 x 17.78 cm) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.