Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 22 (organic cation transporter), member 18 antisense, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004222 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-222 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-222 Supplier Abnova Corporation Supplier No. H00005003A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SLC22A18AS.

Sequence: EVVYSVPDNVPGQNGSRRPLVCKITGKCLSVCSEENAKAGGCSAFPLLLSQLGARMTGREHAHKGPELTTPDSGLPRPPNPALAGFRALAQHSPPLGTSTPSAVLLSAAT

Specifications

Antigen solute carrier family 22 (organic cation transporter), member 18 antisense
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC22A18AS.
Formulation 50% glycerol
Gene SLC22A18AS
Gene Accession No. NM_007105
Gene Alias BWR1B/BWSCR1B/ORCTL2S/SLC22A1LS/p27-BWR1B
Gene Symbols SLC22A18AS
Host Species Mouse
Immunogen SLC22A18AS (NP_009036, 144 a.a. ∼ 253 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5003
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.