Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A3, Mouse, Clone: 2E3, Abnova™

Catalog No. 89021352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-352 Supplier Abnova Corporation Supplier No. H00001811M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC26A3.

The protein encoded by this gene is a transmembrane glycoprotein that transports chloride ions across the cell membrane in exchange for bicarbonate ions. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. The protein is essential for intestinal chloride absorption, and mutations in this gene have been associated with congenital chloride diarrhea. [provided by RefSeq

Sequence: TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDT

Specifications

Antigen SLC26A3
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC26A3.
Formulation PBS with no preservative; pH 7.4
Gene SLC26A3
Gene Accession No. NM_000111
Gene Alias CLD/DRA
Gene Symbols SLC26A3
Host Species Mouse
Immunogen SLC26A3 (NP_000102, 503 a.a. ∼ 600 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1811
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.